missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-toluenesulfonate, 97%

Product Code. 10731061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10731061 100 g Glass bottle 100g
10667142 500 g Glass bottle 500g
10702774 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10731061 Supplier Thermo Scientific Acros Supplier No. 170101000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-48-8 | C8H10O3S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-48-8
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00008417
InChI Key VUQUOGPMUUJORT-UHFFFAOYSA-N
Synonym methyl p-toluenesulfonate, methyl tosylate, methyl p-tosylate, methyl toluene-4-sulfonate, methyl-p-toluenesulfonate, p-toluenesulfonic acid, methyl ester, benzenesulfonic acid, 4-methyl-, methyl ester, methyl toluene-4-sulphonate, p-toluenesulfonic acid methyl ester, p-methylbenzenesulfonate methyl ester
PubChem CID 6645
IUPAC Name methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC

Specifications

Acidity 0.2% max.
Melting Point 24.0°C to 30.0°C
Color White to Yellow
Boiling Point 292.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4SO3CH3
Refractive Index 1.5162 to 1.5182
Quantity 100 g
Beilstein 11, 99
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and benzene
Formula Weight 186.23
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Methyl p-toluenesulfonate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl p-toluenesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.