missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-toluate, 99%

Product Code. 11915001 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
10kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 11915001

Brand: Thermo Scientific Acros 414960100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-75-2 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-75-2
MDL Number MFCD00008441
InChI Key QSSJZLPUHJDYKF-UHFFFAOYSA-N
Synonym methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
PubChem CID 7455
IUPAC Name methyl 4-methylbenzoate
SMILES CC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 32.0°C to 35.0°C
Density 1.0600g/mL
Boiling Point 220.0°C to 223.0°C
Flash Point 90°C
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Molecular Formula C9H10O2
Linear Formula CH3C6H4COOCH3
Quantity 10 kg
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ethanol,ether
Molecular Weight (g/mol) 150.18
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Methyl p-toluate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl p-toluate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.