missing translation for 'onlineSavingsMsg'
Learn More

Methyl oxalyl chloride, 97%

Product Code. 10502354 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
50 mL
Packaging:
Glass bottle
Unit Size:
25mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10502354 50 mL Glass bottle 50mL
10377820 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10502354 Supplier Thermo Scientific Acros Supplier No. 166000500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5781-53-3 | C3H3ClO3 | 122.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5781-53-3
Molecular Formula C3H3ClO3
Molecular Weight (g/mol) 122.51
MDL Number MFCD00000705
InChI Key ZXUQEPZWVQIOJE-UHFFFAOYSA-N
Synonym methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate
PubChem CID 79846
IUPAC Name methyl 2-chloro-2-oxoacetate
SMILES COC(=O)C(=O)Cl

Specifications

Density 1.3300g/mL
Boiling Point 118°C to 120°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCOCO2CH3
Refractive Index 1.4179 to 1.4199
Quantity 50 mL
Beilstein 02, 541
Specific Gravity 1.33
Solubility Information Solubility in water: decomposes
Formula Weight 122.51
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl oxalyl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl oxalyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.