missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Methyl orotate, 97%
£12.25 - £61.00
Chemical Identifiers
CAS | 6153-44-2 |
---|---|
Molecular Formula | C6H6N2O4 |
Molecular Weight (g/mol) | 170.12 |
MDL Number | MFCD00010564 |
InChI Key | UUTDWTOZAWFKFW-UHFFFAOYSA-N |
Synonym | methyl orotate, orotic acid methyl ester, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, unii-41c6ifg4mq, 4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 6-carbomethyoxyuracil, 41c6ifg4mq, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate |
PubChem CID | 80257 |
IUPAC Name | methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate |
SMILES | COC(=O)C1=CC(=O)NC(=O)N1 |
Description
Chemical Identifiers
6153-44-2 | |
170.12 | |
UUTDWTOZAWFKFW-UHFFFAOYSA-N | |
80257 | |
COC(=O)C1=CC(=O)NC(=O)N1 |
C6H6N2O4 | |
MFCD00010564 | |
methyl orotate, orotic acid methyl ester, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, unii-41c6ifg4mq, 4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 6-carbomethyoxyuracil, 41c6ifg4mq, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate | |
methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate |
Specifications
6153-44-2 | |
Yellow | |
MFCD00010564 | |
14,6876 | |
UUTDWTOZAWFKFW-UHFFFAOYSA-N | |
methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate | |
80257 | |
97% | |
Methyl orotate |
240°C to 244°C | |
C6H6N2O4 | |
881031 | |
methyl orotate, orotic acid methyl ester, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, unii-41c6ifg4mq, 4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 6-carbomethyoxyuracil, 41c6ifg4mq, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate | |
COC(=O)C1=CC(=O)NC(=O)N1 | |
170.12 | |
170.12 | |
Powder |