missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange, TRC

Product Code. 30481231
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481231 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481231 Supplier Toronto Research Chemicals Supplier No. TRCM33011350G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl orange
CAS 547-58-0
Molecular Formula C14 H14 N3 O3 S . Na
Quantity 50 g
Synonym Benzenesulfonic acid, 4-[[4-(dimethylamino)phenyl]azo]-, sodium salt (9CI), Benzenesulfonic acid, p-[[p-(dimethylamino)phenyl]azo]-, sodium salt (8CI), Orange III (6CI), 4-Dimethylaminoazobenzene-4'-sulfonic acid sodium salt, Acid Orange 52, Albion Methyl Orange, C.I. 13025, C.I. Acid Orange 52, Diazoben, Eniamethyl Orange, Gold orange, Helianthine, Helianthine B, KCA Methyl Orange, Methyl Orange B, Methyl orange, Orange 3, Sodium 4-(dimethylamino)azobenzene-4'-sulfonate, Sodium 4-[(4-dimethylamino)phenyldiazenyl]benzenesulfonate, Sodium 4-[4-(dimethylamino)phenylazo]benzenesulfonate, Sodium 4'-(dimethylamino)azobenzene-4-sulfonate, Sodium p-[[p-(dimethylamino)phenyl]azo]benzenesulfonate, Sodium p-dimethylaminoazobenzenesulfonate, Tropaeolin D
InChI Formula InChI=1S/C14H15N3O3S.Na/c1-17(2)13-7-3-11(4-8-13)15-16-12-5-9-14(10-6-12)21(18,19)20;/h3-10H,1-2H3,(H,18,19,20);/q;+1/p-1/b16-15+;
SMILES [Na].O=S(=O)(O)C1=CC=C(N=NC2=CC=C(C=C2)N(C)C)C=C1
IUPAC Name sodium;4-[(E)-[4-(dimethylamino)phenyl]azo]benzenesulfonate
Molecular Weight (g/mol) 327.33
Formula Weight 327.0654
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.