missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange Screened Indicator 0.1% Alcoholic Solution, Reagecon
BIOPHARMA

Product Code. 11583122 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11583122 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11583122 Supplier Reagecon Supplier No. MOS05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.

  • Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
  • For manual titrations; indicator titrant or analyte change color and this color change are by far the most important method of end point detection.
  • This high-quality indicator solution facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.

Specifications

Description Accuracy ± 0.1%
Packaging HDPE Bottle GL45
Physical Form Liquid
Concentration 0.1%
Technique Titration, Analyte Color Change
Quantity 500 mL
compliance-icons compliance-icons
Product Identifier
  • SCREENED METHYL ORANGE INDICATOR SOLUTION
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contain: ethyl alcohol, Methyl Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.