missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange, Pure, Indicator Grade, Solid, C.I.13025, Fisher Chemical™

Product Code. 10753611 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10753611 25 g 25g
10192050 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10753611 Supplier Fisher Chemical Supplier No. M/5000/46

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number 7502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 300°C
Formula Weight 327.32g/mol
Packaging Glass Jar
Color Orange
Quantity 25 g
Physical Form Powder
compliance-icons
Product Identifier
  • Methyl Orange
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.