missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Orange, ACS

Product Code. 11393397
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393397 100 g 100g
11383397 5 g 5g
11373397 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11393397 Supplier Thermo Scientific Chemicals Supplier No. 017874.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol

Methyl orange is a pH indicator frequently used in titrations, also used for histological microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number MFCD00007502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
Synonym Acid Orange 52; C.I. 13025
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Tekniske data

Melting Point >300°C
UN Number UN3143
Beilstein 4732884
Merck Index 14,6105
Solubility Information Soluble in 500 parts water; More soluble in hot water
Formula Weight 327.34
Grade ACS
Odor Weak, characteristic
Color Orange
pH 6.5
Quantity 100 g
Physical Form Crystalline
Chemical Name or Material Methyl Orange
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Methyl Orange, ACS
Signalord
  • Danger
Farekategori
  • Acute toxicity Category 3
Faresætning
  • H301-Toxic if swallowed.
Sikkerhedssætning
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplerende oplysninger
  • MIXTURE LIST-Contains : C.I. Acid Orange 52

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.