missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Orange, ACS reagent

Product Code. 10299930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10299930 10 g Glass bottle 10g
10615501 25 g Glass bottle 25g
10724004 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10299930 Supplier Thermo Scientific Chemicals Supplier No. 414660100
  • ONLINE EXCLUSIVE PRICE
    £18.40
    £16.65 / 10g
    Save £1.75
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number MFCD00007502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
Synonym 4-[4-(Dimethylamino)phenylazo]benzenesulfonic acid, sodium salt, Acid Orange 52, C.I. 13025, MO, Helianthise
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300.0°C
Clarity Passes Test
Beilstein 16, 331
Merck Index 15, 6178
Solubility Information Solubility in water: soluble in hot water. Other solubilities: practically insoluble in alcohol
Infrared Spectrum Authentic
Formula Weight 327.32
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 3.2 (pink or red) to pH 4.4 (yellow)
Color Orange
Quantity 10 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Methyl Orange, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Methyl Orange
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.