missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Orange, 0.1% w/v aq. soln.

Product Code. 15491487
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491487 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15491487 Supplier Thermo Scientific Chemicals Supplier No. 038695.AP
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 547-58-0
Odor Odorless
Molecular Formula C14H14N3NaO3S
MDL Number MFCD00007502
Density 0.987
Color Orange
Quantity 500 mL
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
Physical Form Liquid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
Molecular Weight (g/mol) 327.334
PubChem CID 23673835
Formula Weight 327.34
Chemical Name or Material Methyl orange
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.