missing translation for 'onlineSavingsMsg'
Learn More

Methyl oleate, technical

Product Code. 10434315 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10434315 5 kg Plastic drum 5kg
10760672 500 g Glass bottle 500g
10760482 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10434315 Supplier Thermo Scientific Acros Supplier No. 414640050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67762-38-3 | C19H36O2 | 296.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67762-38-3
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.50
MDL Number MFCD00009578
InChI Key QYDYPVFESGNLHU-ZHACJKMWSA-N
Synonym methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112
PubChem CID 5364509
ChEBI CHEBI:27542
IUPAC Name methyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OC

Specifications

Melting Point -20.0°C
Color Yellow to Amber
Density 0.8700g/mL
Boiling Point 186.0°C (9.0 mmHg)
Flash Point 180°C
Infrared Spectrum Authentic
Packaging Plastic drum
Linear Formula CH3(CH2)7CHCH(CH2)7COOCH3
Refractive Index 1.4510 to 1.4580
Quantity 5 kg
Specific Gravity 0.87
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Viscosity 6.829 mPa.s (20°C)
Formula Weight 296.48
Percent Purity ≥55%
Grade Technical
Physical Form Oily Liquid
Chemical Name or Material Methyl oleate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.