missing translation for 'onlineSavingsMsg'
Learn More

Methyl octanoate, 99%

Product Code. 10045710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045710 500 g Glass bottle 500g
10107090 25 g Glass bottle 25g
10381631 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10045710 Supplier Thermo Scientific Acros Supplier No. 414625000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-11-5 | C9H18O2 | 158.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-11-5
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00009551
InChI Key JGHZJRVDZXSNKQ-UHFFFAOYSA-N
Synonym methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
PubChem CID 8091
ChEBI CHEBI:87432
IUPAC Name methyl octanoate
SMILES CCCCCCCC(=O)OC

Specifications

Acidity 1mg KOH/g max.
Color Colorless
Density 0.8780g/mL
Boiling Point 79.0°C (12.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)6COOCH3
Refractive Index 1.4165 to 1.4185
Quantity 500 g
Specific Gravity 0.878
Solubility Information Solubility in water: insoluble
Formula Weight 158.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl octanoate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.