missing translation for 'onlineSavingsMsg'
Learn More

Methyl octanoate, 98+%

Product Code. 11484433
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484433 1000 g 1000g
11494433 50 g 50g
11404443 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484433 Supplier Thermo Scientific Alfa Aesar Supplier No. A10991.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 111-11-5 | C9H18O2 | 158.241 g/mol

Methyl octanoate-ethanol mixtures constitute the biodiesel-bioethanol surrogate fuel and kinetics of its oxidation has been studied experimentally in a jet-stirred reactor. Deoxygenation of methyl octanoate over alumina-supported Pt has been studied in both the vapor phase in a flow reactor and in the liquid phase in a semi-batch reactor. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl octanoate-ethanol mixtures constitute the biodiesel-bioethanol surrogate fuel and kinetics of its oxidation has been studied experimentally in a jet-stirred reactor. Deoxygenation of methyl octanoate over alumina-supported Pt has been studied in both the vapor phase in a flow reactor and in the liquid phase in a semi-batch reactor. It is also used to make other chemicals.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-11-5
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
MDL Number MFCD00009551
InChI Key JGHZJRVDZXSNKQ-UHFFFAOYSA-N
Synonym methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
PubChem CID 8091
ChEBI CHEBI:87432
IUPAC Name methyl octanoate
SMILES CCCCCCCC(=O)OC

Specifications

Melting Point -40°C
Density 0.875
Boiling Point 193°C to 194°C
Flash Point 72°C (161°F)
Refractive Index 1.4175
Quantity 1000 g
Beilstein 1752270
Solubility Information Insoluble in water.
Formula Weight 158.24
Percent Purity 99%
Chemical Name or Material Methyl octanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.