missing translation for 'onlineSavingsMsg'
Learn More

Methyl o-tolylacetate, 98%, Thermo Scientific™

Product Code. p-7045226
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322327 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11322327 Supplier Thermo Scientific Alfa Aesar Supplier No. L09877.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methyl o-tolylacetate is used as pharmaceutical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40851-62-5
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00082780
InChI Key BLEMRRXGTKTJGT-UHFFFAOYSA-N
Synonym methyl o-tolylacetate, methyl 2-2-methylphenyl acetate, methyl 2-o-tolylacetate, methyl 2-o-tolyl acetate, methyl 2-methylphenylacetate, methyl 2-methylphenyl-acetate, methyl 2-methylphenyl acetate, methyl 2-methyl phenyl acetate, 2-methylbenzeneacetic acid methyl ester, 2-methylphenyl acetic acid methyl ester
PubChem CID 7021479
IUPAC Name methyl 2-(2-methylphenyl)acetate
SMILES COC(=O)CC1=CC=CC=C1C

Specifications

Boiling Point 70°C to 71°C (1 mmHg)
Refractive Index 1.511
Quantity 25 g
Beilstein 2208739
Formula Weight 164.21
Percent Purity 98%
Chemical Name or Material Methyl o-tolylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.