missing translation for 'onlineSavingsMsg'
Learn More

Methyl nonanoate, 95%

Product Code. 10117003 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10117003 100 g Glass bottle 100g
10075103 5 g Glass bottle 5g
10340574 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10117003 Supplier Thermo Scientific Acros Supplier No. 385561000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1731-84-6 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1731-84-6
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00009569
InChI Key IJXHLVMUNBOGRR-UHFFFAOYSA-N
Synonym methyl pelargonate, pelargonic acid methyl ester, methyl nonylate, nonanoic acid, methyl ester, methyl n-nonanoate, nonanoic acid methyl ester, unii-7xkd6qoh68, fema no. 2724, methyl ester nonanoic acid, 7xkd6qoh68
PubChem CID 15606
ChEBI CHEBI:44499
IUPAC Name methyl nonanoate
SMILES CCCCCCCCC(=O)OC

Specifications

Melting Point -30°C
Color Colorless
Density 0.8740g/mL
Boiling Point 213°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7C(O)OCH3
Refractive Index 1.4210 to 1.423
Quantity 100 g
Specific Gravity 0.874
Solubility Information Solubility in water: practically insoluble
Formula Weight 172.27
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl nonanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.