missing translation for 'onlineSavingsMsg'
Learn More

Methyl Nicotinate, TRC

Product Code. 30010069
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010069 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30010069 Supplier Toronto Research Chemicals Supplier No. TRCM32324010G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 93-60-7
Molecular Formula C7 H7 N O2
Quantity 10 g
Synonym Methyl Nicotinate, 3-Pyridinecarboxylic acid, methyl ester (9CI, ACI), Nicotinic acid, methyl ester (6CI, 7CI, 8CI), 3-(Carbomethoxy)pyridine, 3-(Methoxycarbonyl)pyridine, m-(Methoxycarbonyl)pyridine, Methyl 3-pyridinecarboxylate, Nicometh, NSC 13126, NSC 403799
InChI Formula InChI=1S/C7H7NO2/c1-10-7(9)6-3-2-4-8-5-6/h2-5H,1H3
SMILES COC(=O)c1cccnc1
IUPAC Name methyl pyridine-3-carboxylate
Molecular Weight (g/mol) 137.14
Formula Weight 137.0477
Chemical Name or Material Methyl nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.