missing translation for 'onlineSavingsMsg'
Learn More

Methyl nicotinate, MedChemExpress

Product Code. 16458796
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458796 Supplier Medchem Express Supplier No. HYB1695

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methyl nicotinate, the methyl ester of Niacin found in alcoholic beverages, that is used as an active ingredient as a rubefacient in over-the-counter topical preparations indicated for muscle and joint pain.

Methyl nicotinate, the methyl ester of Niacin found in alcoholic beverages, that is used as an active ingredient as a rubefacient in over-the-counter topical preparations indicated for muscle and joint pain.

Specifications

Chemical Name or Material Methyl nicotinate
CAS 93-60-7
Color White
Purity Grade Notes Research
Molecular Formula C7H7NO2
Quantity 100 mg
Solubility Information H2O : 100 mg/mL (729.18 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (729.18 mM; Need ultrasonic)
SMILES O=C(C1=CC=CN=C1)OC
Molecular Weight (g/mol) 137.14
Formula Weight 137.14
Percent Purity 99.8%
Grade Research
Physical Form Crystals
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.