missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl N-trimethylsilylcarbamate, 96%

Product Code. 15469059
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15469059

Brand: Alfa Aesar™ L17352.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 66°C to 67°C (15mmHg)
CAS 18147-09-6
Quantity 5 g
Molecular Formula C5H13NO2Si
Linear Formula (CH3)3SiNHCO2CH3
MDL Number MFCD00236515
Beilstein 2350760
Synonym methyl trimethylsilyl carbamate, acmc-20akf5, methyl trimethylsilylcarbamate, trimethylsilyl methyl carbamate, methyl n-trimethylsilyl carbamate, trimethylsilyl carbamic acid methyl ester, carbamic acid, trimethylsilyl-, methyl ester, carbamic acid, trimethylsilyl-,methyl ester 7ci,8ci,9ci
InChI Key JUKSJRIXGQOQTH-UHFFFAOYSA-N
SMILES COC(=O)N[Si](C)(C)C
IUPAC Name methyl N-trimethylsilylcarbamate
Molecular Weight (g/mol) 147.25
PubChem CID 328380
Formula Weight 147.25
Percent Purity 96%
Refractive Index 1.4267
Sensitivity Moisture sensitive
Density 0.963
Chemical Name or Material Methyl N-trimethylsilylcarbamate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.