missing translation for 'onlineSavingsMsg'
Learn More

Methyl Myristate, TRC

Product Code. 30011071
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011071 10 g 10g
17959844 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011071 Supplier Toronto Research Chemicals Supplier No. TRCM22760010G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-10-7
Molecular Formula C15 H30 O2
Molecular Weight (g/mol) 242.4
Synonym Tetradecanoic Acid Methyl Ester, Tetradecanoic acid, methyl ester, Myristic acid, methyl ester (6CI,8CI), Emery 2214, Metholeneat 2495, Methyl myristate, Methyl n-tetradecanoate, Methyl tetradecanoate, NSC 5029, Pastell M 14, Uniphat A50
IUPAC Name methyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17-2/h3-14H2,1-2H3
Formula Weight 242.2246
Chemical Name or Material Methyl myristate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.