missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl myristate, 99%, Thermo Scientific Chemicals
£22.00 - £198.00
Chemical Identifiers
CAS | 124-10-7 |
---|---|
Molecular Formula | C15H30O2 |
Molecular Weight (g/mol) | 242.40 |
MDL Number | MFCD00008983 |
InChI Key | ZAZKJZBWRNNLDS-UHFFFAOYSA-N |
Synonym | methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci |
PubChem CID | 31284 |
IUPAC Name | methyl tetradecanoate |
SMILES | CCCCCCCCCCCCCC(=O)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10581991
|
Thermo Scientific Acros
165010025 |
2.5 mL | Glass bottle |
£22.00
2.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10012020
|
Thermo Scientific Acros
165011000 |
100 mL | Glass bottle |
£90.90
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337390
|
Thermo Scientific Acros
165015000 |
500 mL | Glass bottle |
£198.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-10-7 | |
242.40 | |
ZAZKJZBWRNNLDS-UHFFFAOYSA-N | |
31284 | |
CCCCCCCCCCCCCC(=O)OC |
C15H30O2 | |
MFCD00008983 | |
methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci | |
methyl tetradecanoate |
Specifications
1mg KOH/g max. | |
18.4°C | |
0.8630g/mL | |
>110°C | |
98.5% min. (GC) | |
CH3(CH2)12CO2CH3 | |
MFCD00008983 | |
0.863 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol | |
CCCCCCCCCCCCCC(=O)OC | |
242.40 | |
242.4 | |
Liquid |
124-10-7 | |
Colorless to Yellow | |
323.0°C | |
Authentic | |
C15H30O2 | |
1.4340 to 1.4380 | |
02, II, 326 | |
methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci | |
ZAZKJZBWRNNLDS-UHFFFAOYSA-N | |
methyl tetradecanoate | |
31284 | |
99% | |
Methyl myristate, 99% |
Safety and Handling
missing translation for 'einecsNumber' : 204-680-1
missing translation for 'tsca' : TSCA
RUO – Research Use Only