missing translation for 'onlineSavingsMsg'
Learn More

Methyl methoxyacetate, 99%

Product Code. 10035813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10035813 25 g Glass bottle 25g
10562761 100 g Glass bottle 100g
10042030 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10035813 Supplier Thermo Scientific Acros Supplier No. 165090250
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6290-49-9 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6290-49-9
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00008451
InChI Key QRMHDGWGLNLHMN-UHFFFAOYSA-N
Synonym methyl methoxyacetate, acetic acid, methoxy-, methyl ester, methoxyacetic acid methyl ester, methyl-2-methoxyacetate, methoxyacetic acid, methyl ester, acetic acid, 2-methoxy-, methyl ester, unii-n960nq69lf, methoxy acetic acid methyl ester, methyl metoxyacetate, methyl methoxylacetate
PubChem CID 80507
ChEBI CHEBI:34841
IUPAC Name methyl 2-methoxyacetate
SMILES COCC(=O)OC

Specifications

Color Colorless to Brown
Density 1.0500g/mL
Boiling Point 129°C to 130°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OCH2CO2CH3
Refractive Index 1.3954 to 1.3974
Quantity 25 g
Beilstein 03, 236
Specific Gravity 1.05
Solubility Information Solubility in water: soluble
Formula Weight 104.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl methoxyacetate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl methoxyacetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.