missing translation for 'onlineSavingsMsg'
Learn More

Methyl methoxyacetate, 99%

Product Code. p-7026503
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11438800

Brand: Thermo Scientific Alfa Aesar B22528.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6290-49-9 | C4H8O3 | 104.11 g/mol

Methyl methoxyacetate is used in the preparation of 4-hydroxy-2-mercapto-5-methoxypyrimidine (II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methoxyacetate is used in the preparation of 4-hydroxy-2-mercapto-5-methoxypyrimidine (II).

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6290-49-9
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00008451
InChI Key QRMHDGWGLNLHMN-UHFFFAOYSA-N
Synonym methyl methoxyacetate, acetic acid, methoxy-, methyl ester, methoxyacetic acid methyl ester, methyl-2-methoxyacetate, methoxyacetic acid, methyl ester, acetic acid, 2-methoxy-, methyl ester, unii-n960nq69lf, methoxy acetic acid methyl ester, methyl metoxyacetate, methyl methoxylacetate
PubChem CID 80507
ChEBI CHEBI:34841
IUPAC Name methyl 2-methoxyacetate
SMILES COCC(=O)OC

Specifications

Boiling Point 129°C to 130°C
Flash Point 35°C (95°F)
Quantity 25 g
UN Number UN3272
Beilstein 1742944
Formula Weight 104.11
Percent Purity 99%
Chemical Name or Material Methyl methoxyacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.