missing translation for 'onlineSavingsMsg'
Learn More

Methyl methoxyacetate, 99%

Product Code. 11490727
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490727 500 g 500g
11438800 25 g 25g
11480727 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490727 Supplier Thermo Scientific Alfa Aesar Supplier No. B22528.36
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 6290-49-9 | C4H8O3 | 104.11 g/mol

Methyl methoxyacetate is used in the preparation of 4-hydroxy-2-mercapto-5-methoxypyrimidine (II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methoxyacetate is used in the preparation of 4-hydroxy-2-mercapto-5-methoxypyrimidine (II).

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6290-49-9
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00008451
InChI Key QRMHDGWGLNLHMN-UHFFFAOYSA-N
Synonym methyl methoxyacetate, acetic acid, methoxy-, methyl ester, methoxyacetic acid methyl ester, methyl-2-methoxyacetate, methoxyacetic acid, methyl ester, acetic acid, 2-methoxy-, methyl ester, unii-n960nq69lf, methoxy acetic acid methyl ester, methyl metoxyacetate, methyl methoxylacetate
PubChem CID 80507
ChEBI CHEBI:34841
IUPAC Name methyl 2-methoxyacetate
SMILES COCC(=O)OC

Specifications

Density 1.051
Boiling Point 129°C to 130°C
Flash Point 35°C (95°F)
Refractive Index 1.396
Quantity 500 g
UN Number UN3272
Beilstein 1742944
Solubility Information Soluble in water.
Formula Weight 104.11
Percent Purity 99%
Chemical Name or Material Methyl methoxyacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.