missing translation for 'onlineSavingsMsg'
Learn More

Methyl mercaptoacetate, 95%

Product Code. 11386646
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386646 500 g 500g
11366646 25 g 25g
11376646 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11386646 Supplier Thermo Scientific Alfa Aesar Supplier No. L03783.36
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2365-48-2 | C3H6O2S | 106.14 g/mol

Methyl thioglycolate was used in the preparation of 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene. It is used to prepare methyl thioglycolate and aminoethanethiol conjugated gold nanorods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl thioglycolate was used in the preparation of 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene. It is used to prepare methyl thioglycolate and aminoethanethiol conjugated gold nanorods.

Solubility
Soluble in water. (40 g/L at 20°C)

Notes
Air Sensitive. Store under dry inert gas. Incompatible with oxidizing agents and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2365-48-2
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00004873
InChI Key MKIJJIMOAABWGF-UHFFFAOYSA-N
Synonym methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el
PubChem CID 16907
IUPAC Name methyl 2-sulfanylacetate
SMILES COC(=O)CS

Specifications

Melting Point -3°C
Density 1.163
Boiling Point 148°C to 149°C
Flash Point 56°C (132°F)
Odor Stench
Linear Formula HSCH2COOCH3
Refractive Index 1.466
Quantity 500 g
UN Number UN1228
Beilstein 506259
Sensitivity Air Sensitive
Solubility Information Soluble in water. (40g/L at 20°C)
Formula Weight 106.14
Percent Purity 98%
Chemical Name or Material Methyl mercaptoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.