missing translation for 'onlineSavingsMsg'
Learn More

Methyl m-toluate, 98%

Product Code. 11444253
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444253 50 g 50g
11454253 250 g 250g
11434253 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444253 Supplier Thermo Scientific Alfa Aesar Supplier No. A10893.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99-36-5 | C9H10O2 | 150.177 g/mol

Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl3-methylcyclohexanecarboxylate was prepared by hydrogenation of methyl m-toluate in acetic acid solvent over Adams catalyst. analysis for valproic acid (VA) in serum, for which we use a gas chromatograph equipped with a flame ionization detector, with methyl-m-toluate (MMT) as internal standard.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-36-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008436
InChI Key CPXCDEMFNPKOEF-UHFFFAOYSA-N
Synonym methyl m-toluate, methyl 3-toluate, benzoic acid, 3-methyl-, methyl ester, methyl m-methylbenzoate, meta-toluic acid, methyl ester, 3-methylbenzoic acid methyl ester, methyl-m-methyl benzoate, m-toluic acid, methyl ester, methyl3-methylbenzoate, m-toluic acid methyl ester
PubChem CID 7435
IUPAC Name methyl 3-methylbenzoate
SMILES CC1=CC=CC(=C1)C(=O)OC

Specifications

Density 1.062
Boiling Point 113°C (27 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.516
Quantity 50 g
Beilstein 2043574
Solubility Information Insoluble in water.
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material Methyl m-toluate
compliance-icons
Product Identifier
  • Methyl m-toluate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Methyl m-toluate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.