missing translation for 'onlineSavingsMsg'
Learn More

Methyl Linolenate, TRC

Product Code. 30482085
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482085 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30482085 Supplier Toronto Research Chemicals Supplier No. TRCM315700500MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Linolenic Acid Methyl Ester
CAS 301-00-8
Purity Grade Notes HPLC
Molecular Formula C19 H32 O2
Quantity 500 mg
Synonym 9,12,15-Octadecatrienoic acid','methyl ester','(Z,Z,Z)-, Linolenic acid','methyl ester (6CI,8CI), (Z,Z,Z)-9,12,15-Octadecatrienoic acid methyl ester, 9Z,12Z,15Z-Octadecatrienoic acid methyl ester, Methyl (9Z,12Z,15Z)-9,12,15-octadecatrienoate, Methyl (9Z,12Z,15Z)-octadecatrienoate, Methyl (Z,Z,Z)-9,12,15-octadecatrienoate, Methyl all-cis-9,12,15-octadecatrienoate, Methyl cis,cis,cis-octadeca-9,12,15-trienoate, Methyl linolenate, Methyl α-linolenate, α-Linolenic acid methyl ester
InChI Formula InChI=1S/C19H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h4-5,7-8,10-11H,3,6,9,12-18H2,1-2H3/b5-4-,8-7-,11-10-
SMILES CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC
IUPAC Name methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
Molecular Weight (g/mol) 292.46
Formula Weight 292.2402
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.