missing translation for 'onlineSavingsMsg'
Learn More

Amsacrine hydrochloride, MedChemExpress

Product Code. 16461907
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16461907

Brand: Medchem Express HY13551A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Amsacrine hydrochloride (m-AMSA hydrochloride; acridinyl anisidide hydrochloride) is an inhibitor of topoisomerase II, and acts as an antineoplastic agent which can intercalates into the DNA of tumor cells.

Amsacrine hydrochloride (m-AMSA hydrochloride

Specifications

Chemical Name or Material Amsacrine hydrochloride
CAS 54301-15-4
Color Orange
Purity Grade Notes Research
Molecular Formula C21H20ClN3O3S
Quantity 50 mg
Synonym m-AMSA hydrochloride acridinyl anisidide hydrochloride
Solubility Information DMSO : 62.5 mg/mL (145.38 mM; Need ultrasonic)
SMILES CS(=O)(NC1=CC=C(NC2=C(C=CC=C3)C3=NC4=CC=CC=C42)C(OC)=C1)=O.[H]Cl
Molecular Weight (g/mol) 429.92
Formula Weight 429.92
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.