missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isocyanoacetate, TRC

Product Code. 30475701
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475701 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30475701 Supplier Toronto Research Chemicals Supplier No. TRCM31405010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39687-95-1
Molecular Formula C4H5NO2
Quantity 10 g
Synonym 2-Isocyanoacetic Acid Methyl Ester, Methyl 2-Isocyanoacetate, Methyl alpha-Isocyanoacetate
InChI Formula InChI=1S/C4H5NO2/c1-5-3-4(6)7-2/h3H2,2H3
SMILES COC(=O)C[N+]#[C-]
IUPAC Name methyl 2-isocyanoacetate
Molecular Weight (g/mol) 99.09
Formula Weight 99.032
Chemical Name or Material Methyl Isocyanoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.