missing translation for 'onlineSavingsMsg'
Learn More

Methyl isocyanoacetate, 95%

Product Code. 15424037
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424037 5 g 5g
15414037 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424037 Supplier Thermo Scientific Alfa Aesar Supplier No. H31069.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39687-95-1 | C4H7NO2 | 101.11 g/mol

Methyl isocyanoacetate is used in the copper-catalyzed, diastereoselective synthesis of oxazolines. It is involved in the direct aldol reaction with carbonyl compounds to prepare corresponding oxazoline by using catechol-copper network catalyst. Further, it is also involved in four-component Ugi condensation reaction. It undergoes asymmetric aldol reaction with fluorinated benzaldehyde using gold(I) as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl isocyanoacetate is used in the copper-catalyzed, diastereoselective synthesis of oxazolines. It is involved in the direct aldol reaction with carbonyl compounds to prepare corresponding oxazoline by using catechol-copper network catalyst. Further, it is also involved in four-component Ugi condensation reaction. It undergoes asymmetric aldol reaction with fluorinated benzaldehyde using gold(I) as a catalyst.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Light and moisture sensitive. Incompatible with strong oxidizing agents, strong bases, strong acids, hydrated iron(II) sulfate, heavy metals and aluminum. Store in a cool place. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39687-95-1
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00000006
InChI Key ZRGVTTCCASYOFN-UHFFFAOYSA-N
Synonym methyl isocyanoacetate, isocyanoacetic acid methyl ester, methylisocyanoacetate, isocyano-acetic acid methyl ester, acetic acid,2-isocyano-, methyl ester, methyl-isocyanoacetate, methyl isocyano-acetate, acmc-20akf8, methyl alpha-isocyanoacetate, ksc581o3d
PubChem CID 547815
IUPAC Name methyl 2-isocyanoacetate
SMILES COC(=O)CN=C

Specifications

Density 1.09
Boiling Point 75°C to 76°C (10 mmHg)
Flash Point 84°C (183°F)
Linear Formula CNCH2CO2CH3
Quantity 5 g
UN Number UN2922
Sensitivity Light sensitive
Solubility Information Miscible with organic solvents. Slightly miscible with water.
Formula Weight 99.09
Percent Purity 95%
Chemical Name or Material Methyl isocyanoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.