missing translation for 'onlineSavingsMsg'
Learn More

Methyl isocyanoacetate, 95%, Tech.

Product Code. 10348613 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10348613 25 g Glass bottle 25g
10042940 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10348613 Supplier Thermo Scientific Acros Supplier No. 219000250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39687-95-1 | C4H7NO2 | 101.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39687-95-1
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00000006
InChI Key ZRGVTTCCASYOFN-UHFFFAOYSA-N
Synonym methyl isocyanoacetate, isocyanoacetic acid methyl ester, methylisocyanoacetate, isocyano-acetic acid methyl ester, acetic acid,2-isocyano-, methyl ester, methyl-isocyanoacetate, methyl isocyano-acetate, acmc-20akf8, methyl alpha-isocyanoacetate, ksc581o3d
PubChem CID 547815
IUPAC Name methyl 2-isocyanoacetate
SMILES COC(=O)CN=C

Specifications

Color Yellow to Brown
Density 1.0900g/mL
Boiling Point 75°C to 76°C (10.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CNCH2CO2CH3
Refractive Index 1.4161 to 1.4191
Quantity 25 g
Specific Gravity 1.09
Formula Weight 99.09
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material Methyl isocyanoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.