missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isocyanate, SPEX CertiPrep™

Catalog Number 15296196
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Methyl Isocyanate624-83-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS 624-83-9, 110-54-3
CAS Min % 0.1, 99.9
CAS Max % 0.1, 99.9
Molecular Formula C2H3NO
MDL Number MFCD00002038
InChI Key HAMGRBXTJNITHG-UHFFFAOYSA-N
SMILES CN=C=O
IUPAC Name methylimino(oxo)methane
Molecular Weight (g/mol) 57.05
PubChem CID 12228
ChEBI CHEBI:59059
Packaging 2mL amber prescored glass ampule
Solvent or Matrix Hexane
Quantity 1 mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Chemical Name or Material Methyl Isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl Isocyanate
Signal Word
  • Danger||Warning
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local, regional, national and international regulations.
Supplemental information
  • MIXTURE LIST-Contains : n-hexane, methyl isocyanate

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'