missing translation for 'onlineSavingsMsg'
Learn More

Methyl Isobutyrate, TRC

Product Code. 30467478
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467478 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467478 Supplier Toronto Research Chemicals Supplier No. TRCM31324525G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isobutyric acid methyl ester
CAS 547-63-7
Molecular Formula C5 H10 O2
Quantity 25 g
Synonym Isobutyric acid','methyl ester (6CI,8CI), 2-Methylpropanoic acid methyl ester, 2-Methylpropionic acid methyl ester, Methyl 2,2-dimethylacetate, Methyl 2-methylpropanoate, Methyl 2-methylpropionate, Methyl isobutyrate, NSC 126780
InChI Formula InChI=1S/C5H10O2/c1-4(2)5(6)7-3/h4H,1-3H3
SMILES COC(=O)C(C)C
IUPAC Name methyl 2-methylpropanoate
Molecular Weight (g/mol) 102.13
Formula Weight 102.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.