missing translation for 'onlineSavingsMsg'
Learn More

Methyl isobutyrate, 99%

Product Code. 10143890 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143890 100 g Glass bottle 100g
10153890 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143890 Supplier Thermo Scientific Supplier No. 164601000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 547-63-7 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-63-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00008914
InChI Key BHIWKHZACMWKOJ-UHFFFAOYSA-N
Synonym methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne
PubChem CID 11039
ChEBI CHEBI:73689
IUPAC Name methyl 2-methylpropanoate
SMILES CC(C)C(=O)OC

Specifications

Acidity 1mg KOH/g max.
Melting Point -85°C to 84°C
Color Colorless
Density 0.8900g/mL
Boiling Point 90°C
Flash Point 1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCO2CH3
Refractive Index 1.3790 to 1.384
Quantity 100 g
Beilstein 02, 290
Merck Index 15, 6160
Specific Gravity 0.89
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Viscosity < 30 mPa.s
Formula Weight 102.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl isobutyrate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl isobutyrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.