missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Indole-2,4,5,6,7-d5-3-acetate, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Methyl Indole-2,4,5,6,7-d5-3-acetate |
| CAS | 102415-39-4 |
| Molecular Formula | C11 D5 H6 N O2 |
| Quantity | 0.01 g |
| Synonym | 1 H-Indole-2,4,5,6,7-d5-3-acetic acid, methyl ester (9 CI), 1 H-Indole-2,4,5,6,7-d5-3-acetic acid methyl ester, 1 H-Indole-3-acetic Acid Methyl Ester-D5 (indole-2,4,5,6,7-D5), Methyl Indole-2,4,5,6,7-d5-3-acetate, 2-(2,4,5,6,7-Pentadeuterio-1 H-indol-3-yl)acetic acid methyl ester, Methyl 2-(1 H-indol-3-yl-d5)acetate, Methyl 2-(1 H-indol-3-yl-d5)acetate |
| InChI Formula | InChI=1 S/C11H11NO2/c1-14-11(13)6-8-7-12-10-5-3-2-4-9(8)10/h2-5,7,12 H,6H2,1H3/i2D,3 D,4 D,5 D,7 D |
| SMILES | [2 H]c1[nH]c2c([2 H])c([2 H])c([2 H])c([2 H])c2c1CC(=O)OC |
| IUPAC Name | methyl 2-(2,4,5,6,7-pentadeuterio-1 H-indol-3-yl)acetate |
| Molecular Weight (g/mol) | 194.2414 |
| Formula Weight | 194.1104 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction