missing translation for 'onlineSavingsMsg'
Learn More

Methyl imidazo[1,2-a]pyridine-7-carboxylate, 95%, Thermo Scientific Chemicals

Product Code. 15407507
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15407507 1 g 1g
15498995 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407507 Supplier Thermo Scientific Alfa Aesar Supplier No. H35310.03
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 86718-01-6 | C9H8N2O2 | 176.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86718-01-6
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.18
MDL Number MFCD09910350
InChI Key KYHRVKMXYXBEQN-UHFFFAOYSA-N
Synonym methyl imidazo 1,2-a pyridine-7-carboxylate, imidazo 1,2-a pyridine-7-carboxylic acid methyl ester, 7-methoxycarbonyl imidazo 1,2-a pyridine, methylimidazo 1,2-a pyridine-7-carboxylate, methyl h-imidazo 1,2-a pyridine-7-carboxylate, imidazo 1,2-a pyridine-7-carboxylic acid, methyl ester, pubchem21685, methylimidazoapyridinecarboxylate, methyl imidazo-1,2-a pyridine-7-carboxylate, 7-imidazo 1,2-a pyridinecarboxylic acid methyl ester
PubChem CID 15096964
IUPAC Name methyl imidazo[1,2-a]pyridine-7-carboxylate
SMILES COC(=O)C1=CC2=NC=CN2C=C1

Specifications

Melting Point 133°C to 135°C
Quantity 1 g
Formula Weight 176.18
Percent Purity 95%
Chemical Name or Material Methyl imidazo[1,2-a]pyridine-7-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.