missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrogen phthalate, 97%

Product Code. 11492497
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492497 10 g 10g
11402507 50 g 50g
11412507 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492497 Supplier Thermo Scientific Alfa Aesar Supplier No. A14780.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4376-18-5 | C9H8O4 | 180.16 g/mol

Methyl hydrogen phthalate is used as a phthalate ester metabolite, linked to precocious puberty.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl hydrogen phthalate is used as a phthalate ester metabolite, linked to precocious puberty.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4376-18-5
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002466
InChI Key FNJSWIPFHMKRAT-UHFFFAOYSA-N
Synonym 2-methoxycarbonyl benzoic acid, methyl hydrogen phthalate, monomethyl phthalate, mono-methyl phthalate, phthalic acid monomethyl ester, o-methoxycarbonyl benzoic acid, monomethyl 1,2-benzenedicarboxylate, phthalic acid, monomethyl ester, monomethylphthalate, unii-2stt6d18jr
PubChem CID 20392
IUPAC Name 2-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=CC=C1C(O)=O

Specifications

Melting Point 81°C to 84°C
Quantity 10 g
Beilstein 2048766
Solubility Information Insoluble in water.
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material Methyl hydrogen phthalate
compliance-icons
Product Identifier
  • Methyl hydrogen phthalate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Methyl hydrogen phthalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.