missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl hexanoate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -71.0°C |
Density | 0.8840g/mL |
Boiling Point | 151.0°C |
Flash Point | 42°C |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH3(CH2)4COOCH3 |
Refractive Index | 1.4047 to 1.4067 |
Quantity | 1 kg |
Specific Gravity | 0.884 |
Show More |
Safety and Handling
Product Identifier
- Methyl hexanoate
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'