missing translation for 'onlineSavingsMsg'
Learn More

Methyl hexanoate, 99%

Product Code. 11489642
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489642 1000 g 1000g
11499642 50 g 50g
11409652 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489642 Supplier Thermo Scientific Alfa Aesar Supplier No. A10757.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-70-7 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-70-7
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009510
InChI Key NUKZAGXMHTUAFE-UHFFFAOYSA-N
Synonym methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate
PubChem CID 7824
ChEBI CHEBI:77322
IUPAC Name methyl hexanoate
SMILES CCCCCC(=O)OC

Specifications

Melting Point -71°C
Density 0.885
Boiling Point 150°C to 151°C
Flash Point 45°C (113°F)
Refractive Index 1.405
Quantity 1000 g
UN Number UN3272
Beilstein 1744683
Formula Weight 130.19
Percent Purity 99%
Chemical Name or Material Methyl hexanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.