missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Heptanoate (Heptanoic Acid Methyl Ester), TRC
Specifications
Specifications
| Chemical Name or Material | Methylheptanoate |
| CAS | 106-73-0 |
| Molecular Formula | C8 H16 O2 |
| Quantity | 1 g |
| Synonym | Enanthic methyl ester, Methyl enanthate, Methyl heptanoate, Methyl heptoate, Methyl n-heptanoate, Methyl n-heptylate, Methyl oenanthylate, Radia 30159 |
| InChI Formula | InChI=1S/C8H16O2/c1-3-4-5-6-7-8(9)10-2/h3-7H2,1-2H3 |
| SMILES | CCCCCCC(=O)OC |
| IUPAC Name | methyl heptanoate |
| Molecular Weight (g/mol) | 144.21 |
| Formula Weight | 144.115 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?