missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Heptafluorobutyrate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Methyl Heptafluorobutyrate |
| CAS | 356-24-1 |
| Molecular Formula | C5 H3 F7 O2 |
| Quantity | 1 g |
| Synonym | Butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, methyl ester (ACI), Butanoic acid, heptafluoro-, methyl ester (9CI), Butyric acid, heptafluoro-, methyl ester (6CI, 7CI, 8CI), Methyl 2,2,3,3,4,4,4-heptafluorobutanoate (ACI), 2,2,3,3,4,4,4-Heptafluoro-butyric acid methyl ester, Heptafluorobutyric acid methyl ester, Methyl heptafluorobutanoate, Methyl heptafluorobutyrate, Methyl perfluorobutanoate, Methyl perfluorobutyrate, Perfluorobutanoic acid-methyl ester, Butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, methyl ester (ACI), Butanoic acid, heptafluoro-, methyl ester (9CI), Butyric acid, heptafluoro-, methyl ester (6CI, 7CI, 8CI), Methyl 2,2,3,3,4,4,4-heptafluorobutanoate (ACI), 2,2,3,3,4,4,4-Heptafluoro-butyric acid methyl ester, Heptafluorobutyric acid methyl ester, Methyl heptafluorobutanoate, Methyl heptafluorobutyrate, Methyl perfluorobutanoate, Methyl perfluorobutyrate, Perfluorobutanoic acid-methyl ester |
| InChI Formula | InChI=1S/C5H3F7O2/c1-14-2(13)3(6,7)4(8,9)5(10,11)12/h1H3 |
| SMILES | COC(=O)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC Name | methyl 2,2,3,3,4,4,4-heptafluorobutanoate |
| Molecular Weight (g/mol) | 228.065 |
| Formula Weight | 228.002 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction