missing translation for 'onlineSavingsMsg'
Learn More

Methyl Heptafluorobutyrate, TRC

Product Code. 30481531
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30481531

Brand: Toronto Research Chemicals TRCM0102855G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Heptafluorobutyrate
CAS 356-24-1
Molecular Formula C5 H3 F7 O2
Quantity 5 g
Synonym Butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, methyl ester (ACI), Butanoic acid, heptafluoro-, methyl ester (9CI), Butyric acid, heptafluoro-, methyl ester (6CI, 7CI, 8CI), Methyl 2,2,3,3,4,4,4-heptafluorobutanoate (ACI), 2,2,3,3,4,4,4-Heptafluoro-butyric acid methyl ester, Heptafluorobutyric acid methyl ester, Methyl heptafluorobutanoate, Methyl heptafluorobutyrate, Methyl perfluorobutanoate, Methyl perfluorobutyrate, Perfluorobutanoic acid-methyl ester, Butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, methyl ester (ACI), Butanoic acid, heptafluoro-, methyl ester (9CI), Butyric acid, heptafluoro-, methyl ester (6CI, 7CI, 8CI), Methyl 2,2,3,3,4,4,4-heptafluorobutanoate (ACI), 2,2,3,3,4,4,4-Heptafluoro-butyric acid methyl ester, Heptafluorobutyric acid methyl ester, Methyl heptafluorobutanoate, Methyl heptafluorobutyrate, Methyl perfluorobutanoate, Methyl perfluorobutyrate, Perfluorobutanoic acid-methyl ester
InChI Formula InChI=1S/C5H3F7O2/c1-14-2(13)3(6,7)4(8,9)5(10,11)12/h1H3
SMILES COC(=O)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name methyl 2,2,3,3,4,4,4-heptafluorobutanoate
Molecular Weight (g/mol) 228.065
Formula Weight 228.002
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.