missing translation for 'onlineSavingsMsg'
Learn More

Methyl heptafluorobutyrate, 98%, Thermo Scientific™

Product Code. 11427847
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427847 10 g 10g
11437847 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427847 Supplier Thermo Scientific Alfa Aesar Supplier No. A16522.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 356-24-1
Molecular Formula C5H3F7O2
Molecular Weight (g/mol) 228.066
MDL Number MFCD00000433
InChI Key MRPUVAKBXDBGJQ-UHFFFAOYSA-N
Synonym methyl heptafluorobutyrate, methyl heptafluorobutanoate, methyl perfluorobutyrate, methylheptafluorobutyrate, butanoic acid, heptafluoro-, methyl ester, heptafluorobutyric acid methyl ester, butyric acid, heptafluoro-, methyl ester, butanoic acid, 2,2,3,3,4,4,4-heptafluoro-, methyl ester, heptafluorobutyryl methyl ester, methyl perfluorbutanoate
PubChem CID 67741
IUPAC Name methyl 2,2,3,3,4,4,4-heptafluorobutanoate
SMILES COC(=O)C(C(C(F)(F)F)(F)F)(F)F

Specifications

Melting Point -86°C
Density 1.472
Boiling Point 80°C to 81°C
Flash Point >110°C (230°F)
Odor Recognizable
Refractive Index 1.293
Quantity 10 g
Beilstein 1792131
Formula Weight 228.06
Percent Purity 98%
Chemical Name or Material Methyl heptafluorobutyrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.