missing translation for 'onlineSavingsMsg'
Learn More

Methyl heptadecanoate, Thermo Scientific™

Product Code. p-7227405 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 17730649

Brand: Thermo Scientific Acros 469300010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Alpha Vector METHYLHEPTADECANOATE
Chemical Name or Material Methyl heptadecanoate
CAS 1731-92-6
Melting Point 29°C to 30°C
Color Colorless or White to Yellow
Boiling Point 152°C to 153°C
Flash Point >110°C
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C18 H36 O2
MDL Number 00009001
Quantity 1 g
Synonym Methyl margarate
Molecular Weight (g/mol) 284.48
Physical Form Liquid or Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.