missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Green, zinc chloride salt, pure, certified

Product Code. 10045750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10045750 25 g Glass bottle 25g
11996921 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10045750 Supplier Thermo Scientific Chemicals Supplier No. 414380250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7114-03-6 | C27H35BrClN3·xZnCl2 | 653.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7114-03-6
Molecular Formula C27H35BrClN3·xZnCl2
Molecular Weight (g/mol) 653.23

Specifications

Melting Point 233.0°C
Color Brown-Red or Green
Physical Form Crystalline Powder
Quantity 25 g
Merck Index 14,6082
Synonym C.I. 42590, Ethyl Green, zinc chloride salt
Solubility Information Solubility in water: 70g/L (20°C)
InChI Key ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
SMILES CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]
IUPAC Name zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
PubChem CID 131675247
Formula Weight 653.23
Grade Pure
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Methyl Green, zinc chloride salt
Show More Show Less
compliance-icons
Product Identifier
  • Methyl Green, zinc chloride salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 4-[[4-(Dimethylamino)phenyl][4-(dimethylimi nio)cyclohexa-2,5-dien-1-ylidene]methyl]-Nethyl-N,N-dime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.