missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Green, Pure, C.I.42585, Biological Stain, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| MDL Number | 11879 |
| Formula Weight | 516.95g/mol |
| Packaging | Glass Jar |
| Color | Brown/Red |
| Quantity | 25 g |
| Physical Form | Powder |
Safety and Handling
Product Identifier
- Methyl green
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction