missing translation for 'onlineSavingsMsg'
Learn More

Methyl Green, 25g, MP Biomedicals

Product Code. 11441622 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441622 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11441622 Supplier MP Biomedicals Supplier No. 04806408
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

7114-03-6 DNA stain useful as a tracking dye in acid buffers. Also able to track native DNA in some neutral systems

Specifications

CAS 7114-03-6
Molecular Formula C27H35Cl4N3Zn
Color Green
Quantity 25 g
InChI Key ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
SMILES CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]
IUPAC Name zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
Molecular Weight (g/mol) 608.78
PubChem CID 131675247
Percent Purity 88%
Grade Certified Biological Stain
Chemical Name or Material Methyl Green
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.