missing translation for 'onlineSavingsMsg'
Learn More

Methyl glycolate, 98%

Product Code. 11453478
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453478 5 g 5g
11463478 25 g 25g
11473478 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453478 Supplier Thermo Scientific Alfa Aesar Supplier No. A17870.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-35-5 | C3H6O3 | 90.078 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl glycolate is a chemical reagent used in the synthesis of taxol and rapamycin. It is used in leather tanning and textile dyeing. It acts as a flavoring agent, preservative in food processing, in water treatment industries and as chemical intermediates. Further, it is used in emulsion polymers, ink and paint additive to improve flow properties and impart gloss. It plays an important role in producing triisopropylsilanyloxy-acetic acid methyl ester using imidazole as reagent.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-35-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00004667
InChI Key GSJFXBNYJCXDGI-UHFFFAOYSA-N
Synonym methyl glycolate, methyl hydroxyacetate, acetic acid, hydroxy-, methyl ester, glycolic acid, methyl ester, acetic acid, 2-hydroxy-, methyl ester, glycolic acid methyl ester, unii-7g7r729ej0, hydroxyacetic acid methyl ester, glycolic acid methyl, methyl2-hydroxyethanoate
PubChem CID 66774
IUPAC Name methyl 2-hydroxyacetate
SMILES COC(=O)CO

Specifications

Density 1.184
Boiling Point 150°C to 151°C
Flash Point 67°C (152°F)
Assay Percent Range 98+%
Linear Formula HOCH2CO2CH3
Refractive Index 1.417
Quantity 5 g
Beilstein 1699571
Solubility Information Miscible with water.
Formula Weight 90.08
Percent Purity 98%
Chemical Name or Material Methyl glycolate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.