missing translation for 'onlineSavingsMsg'
Learn More

Methyl glycolate, 98%

Product Code. 10771442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10771442 25 g Glass bottle 25g
11965081 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10771442 Supplier Thermo Scientific Acros Supplier No. 414370250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-35-5 | C3H6O3 | 90.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-35-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
InChI Key GSJFXBNYJCXDGI-UHFFFAOYSA-N
Synonym methyl glycolate, methyl hydroxyacetate, acetic acid, hydroxy-, methyl ester, glycolic acid, methyl ester, acetic acid, 2-hydroxy-, methyl ester, glycolic acid methyl ester, unii-7g7r729ej0, hydroxyacetic acid methyl ester, glycolic acid methyl, methyl2-hydroxyethanoate
PubChem CID 66774
IUPAC Name methyl 2-hydroxyacetate
SMILES COC(=O)CO

Specifications

Melting Point 149.0°C to 151.0°C
Color Colorless
Density 1.1840g/mL
Boiling Point 52.0°C (17.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2COOCH3
Refractive Index 1.4160 to 1.4180
Quantity 25 g
Beilstein 03,236
Specific Gravity 1.184
Formula Weight 90.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl glycolate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.