missing translation for 'onlineSavingsMsg'
Learn More

Methyl gallate, MedChemExpress

Product Code. 16421668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16421668 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16421668 Supplier Medchem Express Supplier No. HYN2010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methyl gallate is a plant phenolic with antioxidant, anticancer, and anti-inflammatory activities. Methyl gallate also shows bacterial inhibition activity. Methyl gallate also has anti-HIV-1 and HIV-1 enzyme inhibitory activities.

Methyl gallate is a plant phenolic with antioxidant, anticancer, and anti-inflammatory activities. Methyl gallate also shows bacterial inhibition activity. Methyl gallate also has anti-HIV-1 and HIV-1 enzyme inhibitory activities.

Specifications

Chemical Name or Material Methyl gallate
CAS 99-24-1
Color White
Purity Grade Notes Research
Molecular Formula C8H8O5
Quantity 5 g
Synonym Gallincin NSC 363001
Solubility Information DMSO : 100 mg/mL (543.04 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC(O)=C(O)C(O)=C1
Molecular Weight (g/mol) 184.15
Formula Weight 184.15
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.