missing translation for 'onlineSavingsMsg'
Learn More

Methyl formate, Reagent Grade, 97%, Honeywell™

Product Code. 15687950 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 1 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15687950 4 x 1 L Glass bottle Pack of 4
15667950 1 L Glass bottle 1 L
15677950 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15687950 Supplier Honeywell Research Chemicals Supplier No. M468374X1L
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O

Specifications

Color Colorless
Density 0.974g/mL at 20°C
Boiling Point 34°C (1013 hPa)
Flash Point −19°C (−2.2°F)
Packaging Glass bottle
Linear Formula HCO2CH3
Quantity 4 x 1 L
UN Number UN1243
Beilstein 1734623
Formula Weight 60.05g/mol
Percent Purity 0.97
Physical Form Liquid
Chemical Name or Material Methyl formate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.